Learn More
Chemglass Life Sciences Funnel, Heavy Duty, 30mL, 54mm Diameter
SDP
Catalog No. 50872149
Encompass_Preferred
Funnel, Heavy Duty, 30mL, 54mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)
Catalog No. 50-872-149 Supplier Chemglass Life Sciences Supplier No. CG176801
Add to Cart
Add to Cart
Funnel, Heavy Duty, 30mL, 54mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)