Learn More
Chemglass Life Sciences Funnel, Heavy Duty, 480mL, 152.4mm Diameter
SDP
Catalog No. 50872152
Encompass_Preferred
Funnel, Heavy Duty, 480mL, 152.4mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)
Catalog No. 50-872-152 Supplier Chemglass Life Sciences Supplier No. CG176804
Funnel, Heavy Duty, 480mL, 152.4mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)