Learn More
Chemglass Life Sciences Funnel, Heavy Duty, 720mL, 184.2mm Diameter
SDP
Supplier:  Chemglass Life Sciences CG176805
Encompass_Preferred
Funnel, Heavy Duty, 720mL, 184.2mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)
Catalog No. 50-872-153