Learn More
Chemglass Life Sciences Funnel, Heavy Duty, 90mL, 76.2mm Diameter
SDP
Catalog No. 50872150
Encompass_Preferred
Funnel, Heavy Duty, 90mL, 76.2mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)
Catalog No. 50-872-150 Supplier Chemglass Life Sciences Supplier No. CG176802
Funnel, Heavy Duty, 90mL, 76.2mm Diameter. Sturdy Polypropylene funnels with ridges to prevent vapor lock. Highly recommended for continuous laboratory work. Steam autoclavable at 121°C (250°F)