Learn More
Synthware™ Hirsch Filter Funnel
GREENER_CHOICESDP
Catalog No. 31502124 Shop All Kemtech Products
Change view
Click to view available options
:
Hirsch Filter Funnel
Catalog No.
31-502-124 Hirsch Filter Funnel
1 options
Catalog No. 31-502-124 Supplier Synthware™ Supplier No. F650010C
Only null left
Add to Cart
Add to Cart
For all microscale application with 14/10 joint
  • With 10mm O.D. fritted disc of coarse porosity
  • For all microscale application with 14/10 joints
  • Capacity above disc: approximately 7mL
Specifications
Type Hirsch Filter Funnel
Volume (Metric) 7 mL