Learn More
Ace Glass, Inc. Funnel, addition, jacketed, 500mL, 24/40 joints, 4mm PTFE stopcock
SDP
Catalog No. 50974745
Encompass_Preferred
500mL graduated addition funnel with outside jacket for cooling and/or heating. Jacket extends from just below shoulder of vessel down to the bottom tube just below the stopcock. Stopcock is a 4mm bore PTFE plug. Bottom drip tube extends to the edge of the lower, 24/40 inner standard taper joint. Top outer reinforced joint is also 24/40. Side hose barbs are size D for 3/8 I.D. tubing.
Catalog No. 50-974-745 Supplier Ace Glass, Inc. Supplier No. 727807
500mL graduated addition funnel with outside jacket for cooling and/or heating. Jacket extends from just below shoulder of vessel down to the bottom tube just below the stopcock. Stopcock is a 4mm bore PTFE plug. Bottom drip tube extends to the edge of the lower, 24/40 inner standard taper joint. Top outer reinforced joint is also 24/40. Side hose barbs are size D for 3/8 I.D. tubing.