Learn More
Chemglass Life Sciences Funnel, Micro Hirsch, Minum-Ware, Coarse Porosity, Complete
SDP
Catalog No. 501532684
Encompass_Preferred
Funnel, Micro Hirsch, Minum-Ware, Coarse Porosity, Complete. Microscale vertically adjustable Hirsch funnel for use with MINUM-WARE flasks or conical reaction vials having 14/10 joints. Funnel has a stem O.D. of 1/4in and a length of 4in. Disc diameter is 10mm and is available in coarse or medium porosity. Complete item consists of a MW-33 adapter and funnel.
Catalog No. 50-153-2684 Supplier Chemglass Life Sciences Supplier No. MW5302
Funnel, Micro Hirsch, Minum-Ware, Coarse Porosity, Complete. Microscale vertically adjustable Hirsch funnel for use with MINUM-WARE flasks or conical reaction vials having 14/10 joints. Funnel has a stem O.D. of 1/4in and a length of 4in. Disc diameter is 10mm and is available in coarse or medium porosity. Complete item consists of a MW-33 adapter and funnel.