Learn More
Hach Company Funnel, Micro
Catalog No. 501996265 Shop All Hach Company Products
Encompass
Ideal for micro work, filling burets, manometers, and small bottles. Steam autoclavable at 250°F (121°C).
Catalog No. 50-199-6265 Supplier Hach Company Supplier No. 2584335
Ideal for micro work, filling burets, manometers, and small bottles. Steam autoclavable at 250°F (121°C).