Learn More
Chemglass Life Sciences 100mm Powder Funnel, Offset, 24/40 Inner Joint, C to C Offset is 40mm
SDP
Catalog No. CG172203
Encompass_Preferred
100mm Powder Funnel, Offset, 24/40 Inner Joint, C to C Offset is 40mm. Funnel has a lower stem made with an offset to permit use with multi-neck flasks.
Catalog No. CG172203 Supplier Chemglass Life Sciences Supplier No. CG172203
100mm Powder Funnel, Offset, 24/40 Inner Joint, C to C Offset is 40mm. Funnel has a lower stem made with an offset to permit use with multi-neck flasks.