Learn More
Chemglass Life Sciences Funnel Only, Medium, Filter Apparatus
SDP
Catalog No. CG140405
Encompass_Preferred
FUNNEL ONLY. Useful for direct filtration applications of small volumes under vacuum. See CG140401, CG140402 & CG140403 for complete apparatus.
Catalog No. CG140405 Supplier Chemglass Life Sciences Supplier No. CG140405
FUNNEL ONLY. Useful for direct filtration applications of small volumes under vacuum. See CG140401, CG140402 & CG140403 for complete apparatus.