Learn More
Chemglass Life Sciences Minum-Ware, Micro Hirsh Funnel Only, Medium, 14/10
SDP
Catalog No. 501532685
Encompass_Preferred
Minum-Ware, Micro Hirsh Funnel Only, Medium, 14/10. Microscale vertically adjustable Hirsch funnel for use with MINUM-WARE flasks or conical reaction vials having 14/10 joints. Funnel has a stem O.D. of 1/4in and a length of 4in. Disc diameter is 10mm and is available in coarse or medium porosity. Complete item consists of a MW-33 adapter and funnel.
Catalog No. 50-153-2685 Supplier Chemglass Life Sciences Supplier No. MW5304
Minum-Ware, Micro Hirsh Funnel Only, Medium, 14/10. Microscale vertically adjustable Hirsch funnel for use with MINUM-WARE flasks or conical reaction vials having 14/10 joints. Funnel has a stem O.D. of 1/4in and a length of 4in. Disc diameter is 10mm and is available in coarse or medium porosity. Complete item consists of a MW-33 adapter and funnel.