Learn More
Cole-Parmer BrandTech Large industrial-size polypropylene funnel, 2 L
Catalog No. NC9935032 Shop All Cole-Parmer Products
Encompass_Preferred
Low density polyethylene (LDPE) funnels feature external ribbing for air release and have a handle tab for easy lifting. Polypropylene and high density polypropylene (HDPE) funnels are translucent and show content levels. Polypropylene funnels are autoclavahle.
Catalog No. NC9935032 Supplier Cole-Parmer Supplier No. 0612130
Low density polyethylene (LDPE) funnels feature external ribbing for air release and have a handle tab for easy lifting. Polypropylene and high density polypropylene (HDPE) funnels are translucent and show content levels. Polypropylene funnels are autoclavahle.