Learn More
Chemglass Life Sciences 100mm Powder Funnel, Offset, 29/42 Inner Joint, C to C Offset is 45mm
SDP
Catalog No. 501531217
Encompass_Preferred
100mm Powder Funnel, Offset, 29/42 Inner Joint, C to C Offset is 45mm. Funnel has a lower stem made with an offset to permit use with multi-neck flasks.
Catalog No. 50-153-1217 Supplier Chemglass Life Sciences Supplier No. CG172204
100mm Powder Funnel, Offset, 29/42 Inner Joint, C to C Offset is 45mm. Funnel has a lower stem made with an offset to permit use with multi-neck flasks.