Learn More
Chemglass Life Sciences 100mm Powder Funnel, 29/42 Inner Joint
SDP
Catalog No. 501531216
Encompass_Preferred
100mm Powder Funnel, 29/42 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60 side angle and a standard taper inner joint.
Catalog No. 50-153-1216 Supplier Chemglass Life Sciences Supplier No. CG172003
100mm Powder Funnel, 29/42 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60 side angle and a standard taper inner joint.