Learn More
Chemglass Life Sciences 150mm Powder Funnel, Modified, 24/40 Inner Joint, 45mm Flat Sided
SDP
Catalog No. CG172105
Encompass_Preferred
150mm Powder Funnel, Modified, 24/40 Inner Joint, 45mm Flat Sided. Similar to CG-1720, but with one side flattened to allow funnel to be used in multi-neck flasks where space is limited. Flat side dimension is the distance between the center of the standard taper inner joint and the flattened side of the funnel. Funnels have 60º angled sides with a standard taper inner joint.
Catalog No. CG172105 Supplier Chemglass Life Sciences Supplier No. CG172105
150mm Powder Funnel, Modified, 24/40 Inner Joint, 45mm Flat Sided. Similar to CG-1720, but with one side flattened to allow funnel to be used in multi-neck flasks where space is limited. Flat side dimension is the distance between the center of the standard taper inner joint and the flattened side of the funnel. Funnels have 60º angled sides with a standard taper inner joint.