Learn More
Chemglass Life Sciences 150mm Powder Funnel, 45/50 Inner Joint
SDP
Catalog No. CG172006
Encompass_Preferred
150mm Powder Funnel, 45/50 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60º side angle and a standard taper inner joint.
Catalog No. CG172006 Supplier Chemglass Life Sciences Supplier No. CG172006
150mm Powder Funnel, 45/50 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60º side angle and a standard taper inner joint.