Learn More
Chemglass Life Sciences Funnel, Powder, 50mm, 14/20 Outer
SDP
Catalog No. 501531234
Encompass_Preferred
Funnel, Powder, 50mm, 14/20 Outer. Powder funnel is useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60 side angle and a standard taper outer joint.
Catalog No. 50-153-1234 Supplier Chemglass Life Sciences Supplier No. CG176202
Funnel, Powder, 50mm, 14/20 Outer. Powder funnel is useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60 side angle and a standard taper outer joint.