Learn More
Ace Glass, Inc. Funnel, powder, 60mm Duran flange bottom, wide mouth top
SDP
Catalog No. 50974726
Encompass_Preferred
Borosilicate glass Powder Funnel with a 60mm Duran flange bottom and a wide-mouth top. The flange matches side ports on ACE spherical pilot plant reactor heads. The funnel is slightly angled to allow for easy pouring of material into large scale reactors. Uses a stainless steel quick-release clamp for connection to the reactor head.
Catalog No. 50-974-726 Supplier Ace Glass, Inc. Supplier No. 725206
Borosilicate glass Powder Funnel with a 60mm Duran flange bottom and a wide-mouth top. The flange matches side ports on ACE spherical pilot plant reactor heads. The funnel is slightly angled to allow for easy pouring of material into large scale reactors. Uses a stainless steel quick-release clamp for connection to the reactor head.