Learn More
Hach Company Funnel-Powder-65 mm Top ID
Catalog No. 5000257122 Shop All Hach Company Products
Encompass
Polypropylene funnel for easy transfer of powders into narrow-necked flasks tubes or vials
Catalog No. 50-002-57122 Supplier Hach Company Supplier No. 2264467
Polypropylene funnel for easy transfer of powders into narrow-necked flasks tubes or vials