Learn More
Chemglass Life Sciences Funnel, Powder, 75mm, 24/40 Outer
SDP
Catalog No. 501531235
Encompass_Preferred
Funnel, Powder, 75mm, 24/40 Outer. Powder funnel is useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60 side angle and a standard taper outer joint.
Catalog No. 50-153-1235 Supplier Chemglass Life Sciences Supplier No. CG176204
Funnel, Powder, 75mm, 24/40 Outer. Powder funnel is useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60 side angle and a standard taper outer joint.