Learn More
Chemglass Life Sciences Funnel, Powder, PTFE, 52mm Diameter, Stem 16mm, 84mm Height
SDP
Catalog No. 50872142
Encompass_Preferred
PTFE powder funnel is totally chemically inert with super smooth non-stick internal finish.Virgin grade PTFE conforming to FDA and USP Class VIChemical Resistance is virtually total.Temperature Range, -200°C to 260°C (-328°F to 500°F).Shelf life is considered indefinite within the confines of normal use and storage conditions.Autoclavable
Catalog No. 50-872-142 Supplier Chemglass Life Sciences Supplier No. CG176402
PTFE powder funnel is totally chemically inert with super smooth non-stick internal finish.Virgin grade PTFE conforming to FDA and USP Class VIChemical Resistance is virtually total.Temperature Range, -200°C to 260°C (-328°F to 500°F).Shelf life is considered indefinite within the confines of normal use and storage conditions.Autoclavable