Learn More
Wilmad Labglass FUNNEL PWDR 150MM 45/50
Catalog No. 50805545
Encompass_Preferred
Funnel Powder. Approximate Top Diameter: 150mm; Joint Size: [ST] 45/50 Inner. For introduction of liquids or powders into containers having the same size standard taper joint.
Catalog No. 50-805-545 Supplier Wilmad Labglass Supplier No. LG8300110
Funnel Powder. Approximate Top Diameter: 150mm; Joint Size: [ST] 45/50 Inner. For introduction of liquids or powders into containers having the same size standard taper joint.