Learn More
California Pacific Lab & Consulting FUNNEL SAFETY HAZARDOUS
SDP
Supplier:  California Pacific Lab & Consulting EF8FS70
Encompass_Preferred
Eco funnel, w/fS-70 screw cap, 8 in., Made of HDPE and has an easy use lid, gasket, positive latch, filter, and an 11 ft. stem to effectively reduce chemical evaporation of hazardous solvents in the lab, 1750mL
Catalog No. 50-590-36