Learn More
California Pacific Lab & Consulting FUNNEL SAFETY HAZARDOUS
SDP
Catalog No. 5059020
Encompass_Preferred
System, Funnel; California Pacific; ECO; Made of HDPE; 8 in.; 2 in. NPT fine thread; Capacity: 2000mL; Suitable for 15, 30, 55 gal. drums; Easy use lid, gasket, and positive latch to close lid; Interior filter; 11 in. stem
Catalog No. 50-590-20 Supplier California Pacific Lab & Consulting Supplier No. EF47161P
System, Funnel; California Pacific; ECO; Made of HDPE; 8 in.; 2 in. NPT fine thread; Capacity: 2000mL; Suitable for 15, 30, 55 gal. drums; Easy use lid, gasket, and positive latch to close lid; Interior filter; 11 in. stem