Learn More
California Pacific Lab & Consulting FUNNEL SAFETY HAZARDOUS
SDP
Catalog No. 5059032
Encompass_Preferred
Funnel, HDPE; California Pacific; ECO; Made of HDPE; Capacity: 2000mL; 8 in.; Cap size: 83mm; Suitable for 10L carboy; Easy use lid, gasket, and positive latch; Interior filter; 11 in. stem
Catalog No. 50-590-32 Supplier California Pacific Lab & Consulting Supplier No. EF883B
Funnel, HDPE; California Pacific; ECO; Made of HDPE; Capacity: 2000mL; 8 in.; Cap size: 83mm; Suitable for 10L carboy; Easy use lid, gasket, and positive latch; Interior filter; 11 in. stem