Learn More
California Pacific Lab & Consulting FUNNEL SAFETY HAZARDOUS
SDP
Catalog No. 5059002
Encompass_Preferred
Funnel, HDPE; California Pacific; ECO; Capacity: 2000mL; 8 in.; Cap size: 70mm; Suitable for 20L carboy; Easy use lid, gasket, and positive latch; Interior filter; 11 in. stem
Catalog No. 50-590-02 Supplier California Pacific Lab & Consulting Supplier No. EF30020
Funnel, HDPE; California Pacific; ECO; Capacity: 2000mL; 8 in.; Cap size: 70mm; Suitable for 20L carboy; Easy use lid, gasket, and positive latch; Interior filter; 11 in. stem