Learn More
Chemglass Life Sciences 10mL Separatory Funnel, Squibb, 14/20 Joint Size
SDP
Catalog No. CG174301
Encompass_Preferred
10mL Separatory Funnel, Squibb, 14/20 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.
Catalog No. CG174301 Supplier Chemglass Life Sciences Supplier No. CG174301
10mL Separatory Funnel, Squibb, 14/20 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.