Learn More
Chemglass Life Sciences 25mL Separatory Funnel, Squibb, 14/20 Joint Size
SDP
Catalog No. CG174303
Encompass_Preferred
25mL Separatory Funnel, Squibb, 14/20 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.
Catalog No. CG174303 Supplier Chemglass Life Sciences Supplier No. CG174303
25mL Separatory Funnel, Squibb, 14/20 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.