Learn More
Chemglass Life Sciences Funnel, Sep, European, 1000mL, 4mm PTFE Stpk, 24/40
SDP
Catalog No. 50873307
Encompass_Preferred
Funnel, Sep, European, 1000mL, 4mm PTFE Stpk, 24/40. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.
Catalog No. 50-873-307 Supplier Chemglass Life Sciences Supplier No. CG174506
Funnel, Sep, European, 1000mL, 4mm PTFE Stpk, 24/40. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.