Learn More
Chemglass Life Sciences Funnel, Sep, European, 125mL, 2mm PTFE Stpk, 24/40
SDP
Catalog No. 501311096
Encompass_Preferred
Funnel, Sep, European, 125mL, 2mm PTFE Stpk, 24/40. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.
Catalog No. 50-131-1096 Supplier Chemglass Life Sciences Supplier No. CG174501
Add to Cart
Add to Cart
Funnel, Sep, European, 125mL, 2mm PTFE Stpk, 24/40. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.