Learn More
Chemglass Life Sciences Funnel, Sep, European, 250mL, 4mm PTFE Stpk, 24/40
SDP
Catalog No. 501311097
Encompass_Preferred
Funnel, Sep, European, 250mL, 4mm PTFE Stpk, 24/40. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.
Catalog No. 50-131-1097 Supplier Chemglass Life Sciences Supplier No. CG174502
Funnel, Sep, European, 250mL, 4mm PTFE Stpk, 24/40. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.