Learn More
Chemglass Life Sciences Funnel, Sep, European, 2000mL, 6mm PTFE Stpk, 45/50
SDP
Catalog No. 50873308
Encompass_Preferred
Funnel, Sep, European, 2000mL, 6mm PTFE Stpk, 45/50. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.
Catalog No. 50-873-308 Supplier Chemglass Life Sciences Supplier No. CG174507
Funnel, Sep, European, 2000mL, 6mm PTFE Stpk, 45/50. European squibb style separatory funnel with a PTFE stopcock. Supplied complete with a glass stopper of the listed size. For replacement stoppers see CG-3000.