Learn More
Wilmad Labglass FUNNEL SEP SQB 1000ML 29/42
Catalog No. 50806420
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 1000mL. Lower Joint Size: [ST] 29/42. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with PTFE Stopcock and lower standard taper inner joint with drip tip.
Catalog No. 50-806-420 Supplier Wilmad Labglass Supplier No. LG8431T108
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 1000mL. Lower Joint Size: [ST] 29/42. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with PTFE Stopcock and lower standard taper inner joint with drip tip.