Learn More
Wilmad Labglass FUNNEL SEP SQB GLS S/C 125ML
Supplier:  Wilmad Labglass LG8420104
Encompass_Preferred
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 125mL. Stopcock Size: 2. Stopper Size: 22. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.
Catalog No. 50-806-444