Learn More
Wilmad Labglass FUNNEL SEP SQB GLS S/C 2000ML
Catalog No. 50805601
Encompass_Preferred
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 2000mL. Stopcock Size: 6. Stopper Size: 38. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.
Catalog No. 50-805-601 Supplier Wilmad Labglass Supplier No. LG8420112
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 2000mL. Stopcock Size: 6. Stopper Size: 38. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.