Learn More
Wilmad Labglass FUNNEL SEP SQB GLS S/C 250ML
Catalog No. 50803325
Encompass_Preferred
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 250mL. Stopcock Size: 4. Stopper Size: 22. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.
Catalog No. 50-803-325 Supplier Wilmad Labglass Supplier No. LG8420106
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 250mL. Stopcock Size: 4. Stopper Size: 22. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.