Learn More
Wilmad Labglass FUNNEL SEP SQB GLS S/C 4000ML
Supplier:  Wilmad Labglass LG8420116
Encompass_Preferred
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 4000mL. Stopcock Size: 8. Stopper Size: 38. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.
Catalog No. 50-806-294