Learn More
Wilmad Labglass FUNNEL SEP SQB GLS S/C 500ML
Catalog No. 50803832
Encompass_Preferred
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 500mL. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.
Catalog No. 50-803-832 Supplier Wilmad Labglass Supplier No. LG8420108
Separatory Funnel, Squibb, Glass Stopcock. Capacity: 500mL. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with a glass Stopcock. Supplied complete with glass Pennyhead flask length Stopper of the size listed.