Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 125ML
Catalog No. 50803326
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 125mL. Stopcock Size: 2. Stopper Size: 22. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.
Catalog No. 50-803-326 Supplier Wilmad Labglass Supplier No. LG8421T104
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 125mL. Stopcock Size: 2. Stopper Size: 22. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.