Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 250ML
Catalog No. 50803861
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 250mL. Stopcock Size: 4. Stopper Size: 22. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.
Catalog No. 50-803-861 Supplier Wilmad Labglass Supplier No. LG8421T106
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 250mL. Stopcock Size: 4. Stopper Size: 22. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.