Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 500ML
Catalog No. 50803802
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 500mL. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.
Catalog No. 50-803-802 Supplier Wilmad Labglass Supplier No. LG8421T108
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 500mL. Stopcock Size: 4. Stopper Size: 27. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.