Learn More
Wilmad Labglass FUNNEL SEP SQB PTFE S/C 60ML
Catalog No. 50807529
Encompass_Preferred
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 60mL. Stopcock Size: 2. Stopper Size: 16. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.
Catalog No. 50-807-529 Supplier Wilmad Labglass Supplier No. LG8421T102
Separatory Funnel, Squibb, PTFE Stopcock. Capacity: 60mL. Stopcock Size: 2. Stopper Size: 16. Squibb separatory funnel with polytetrafluoroethylene (PTFE) Stopcock. Supplied complete with glass Pennyhead flask length Stopper.