Learn More
Chemglass Life Sciences 1000mL Separatory Funnel, Squibb, #27 Outer Stopper Neck, 4mm Glass Stpk
SDP
Catalog No. CG174006
Encompass_Preferred
1000mL Separatory Funnel, Squibb, #27 Outer Stopper Neck, 4mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174006 Supplier Chemglass Life Sciences Supplier No. CG174006
1000mL Separatory Funnel, Squibb, #27 Outer Stopper Neck, 4mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.