Learn More
Chemglass Life Sciences Funnel, Sep, Squibb, 250mL, 4mm PTFE Stpk, #22 Stopper, PE Stopper
SDP
Catalog No. 50873312
Encompass_Preferred
Funnel, Sep, Squibb, 250mL, 4mm PTFE Stpk, #22 Stopper, PE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PE stopper of the listed size. For replacement stoppers see CG-8740.
Catalog No. 50-873-312 Supplier Chemglass Life Sciences Supplier No. CG174704
Funnel, Sep, Squibb, 250mL, 4mm PTFE Stpk, #22 Stopper, PE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PE stopper of the listed size. For replacement stoppers see CG-8740.