Learn More
Chemglass Life Sciences Funnel, Sep, Squibb, 2L, 6mm PTFE Stpk, #38 Stopper, PE Stopper
SDP
Catalog No. 50873315
Encompass_Preferred
Funnel, Sep, Squibb, 2L, 6mm PTFE Stpk, #38 Stopper, PE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PE stopper of the listed size. For replacement stoppers see CG-8740.
Catalog No. 50-873-315 Supplier Chemglass Life Sciences Supplier No. CG174707
Funnel, Sep, Squibb, 2L, 6mm PTFE Stpk, #38 Stopper, PE Stopper. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length PE stopper of the listed size. For replacement stoppers see CG-8740.