Learn More
Chemglass Life Sciences 125mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 2mm Glass Stpk
SDP
Catalog No. CG174003
Encompass_Preferred
125mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 2mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174003 Supplier Chemglass Life Sciences Supplier No. CG174003
125mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 2mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.