Learn More
Chemglass Life Sciences 500mL Separatory Funnel, Squibb, #27 Outer Stopper Neck, 4mm Glass Stpk
SDP
Catalog No. CG174005
Encompass_Preferred
500mL Separatory Funnel, Squibb, #27 Outer Stopper Neck, 4mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174005 Supplier Chemglass Life Sciences Supplier No. CG174005
500mL Separatory Funnel, Squibb, #27 Outer Stopper Neck, 4mm Glass Stpk. Squibb style separatory funnel with a glass stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.