Learn More
Chemglass Life Sciences 2000mL Separatory Funnel, Squibb, #38 Outer Stopper Neck, 6mm PTFE Stpk
SDP
Catalog No. CG174207
Encompass_Preferred
2000mL Separatory Funnel, Squibb, #38 Outer Stopper Neck, 6mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174207 Supplier Chemglass Life Sciences Supplier No. CG174207
2000mL Separatory Funnel, Squibb, #38 Outer Stopper Neck, 6mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.