Learn More
Chemglass Life Sciences 250mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 4mm PTFE Stpk
SDP
Catalog No. CG174204
Encompass_Preferred
250mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 4mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174204 Supplier Chemglass Life Sciences Supplier No. CG174204
250mL Separatory Funnel, Squibb, #22 Outer Stopper Neck, 4mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.