Learn More
Chemglass Life Sciences 30mL Separatory Funnel, Squibb, #13 Outer Stopper Neck, 2mm PTFE Stpk
SDP
Catalog No. CG174201
Encompass_Preferred
30mL Separatory Funnel, Squibb, #13 Outer Stopper Neck, 2mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.
Catalog No. CG174201 Supplier Chemglass Life Sciences Supplier No. CG174201
30mL Separatory Funnel, Squibb, #13 Outer Stopper Neck, 2mm PTFE Stpk. Squibb style separatory funnel with a PTFE stopcock. Supplied complete with a flask length stopper of the listed size. For replacement stoppers see CG-3018.